Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008881.2 | NP_032907.1 | 10, 11 | 2586 | ||
GenBank mRNA | AK220460.1 | - | 7, 8 | 826 | ||
BC138023.1 | - | 9, 10 | 2378 | 8861042 | ||
D86948.1 | - | 10, 11 | 2570 |