Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025570.1 | NP_001020741.1 | 2 | 1315 | ||
NM_011127.2 | NP_035257.1 | 2 | 1315 | |||
NM_175686.3 | NP_783617.2 | 2 | 1315 | |||
GenBank mRNA | AB221686.1 | - | 2 | 300 | ||
AK008428.2 | - | 3 | 145 | |||
AK019503.1 | - | 2 | 126 | |||
AK031075.1 | - | 2 | 281 | |||
AK032773.1 | - | 2 | 882 | |||
AK084387.1 | - | 2 | 1333 | |||
AK089006.1 | - | 2 | 1147 | |||
AK165914.1 | - | 2 | 88 | |||
BC092372.1 | - | 2 | 664 | 30533016 | ||
L06502.1 | - | 2 | 480 | |||
U03873.1 | - | 2 | 1321 | |||
X59725.1 | - | 2 | 441 |