Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017462.2 | NP_059490.2 | 5 | 1460 | ||
GenBank mRNA | AK133584.1 | - | 5 | 1386 | ||
AK148091.1 | - | 5 | 1422 | |||
AK219254.1 | - | 2 | 180 | |||
BC061472.1 | - | 5 | 1374 | 6827238 | ||
BC080291.1 | - | 5 | 1367 | |||
U53584.1 | - | 5 | 1380 |