Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002177.2 | NP_002168.1 | 1 | 830 | ||
GenBank mRNA | A12140.1 | - | 1 | 1074 | ||
AY780805.1 | - | 1 | 430 | |||
BC069095.1 | - | 1 | 503 | 7216909 | ||
BC117290.1 | - | 1 | 536 | 40125841 | ||
BC117292.1 | - | 1 | 536 | 40125843 | ||
X02669.1 | - | 1 | 437 |