Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163557.1 | NP_001157029.1 | 20 | 2094 | ||
NM_008905.2 | NP_032931.2 | 21 | 2363 | |||
GenBank mRNA | AK012152.1 | - | 17 | 1753 | ||
AK166921.1 | - | 11 | 1231 | |||
BC003966.1 | - | 6 | 663 | |||
BC055935.1 | - | 10 | 964 | |||
BC100346.1 | - | 20 | 2094 | 30936010 | ||
U79024.1 | - | 21 | 2148 |