Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159496.1 | NP_001152968.1 | 2 | 854 | ||
NM_001159497.1 | NP_001152969.1 | 2 | 607 | |||
NM_001159498.1 | NP_001152970.1 | 2 | 530 | |||
NM_011151.2 | NP_035281.1 | 2 | 854 | |||
GenBank mRNA | AJ271833.1 | - | 2 | 832 | ||
AJ271836.1 | - | 2 | 840 | |||
AK050170.1 | - | 2 | 855 | |||
AK076923.1 | - | 2 | 610 | |||
AK077136.1 | - | 3 | 601 | |||
AK136077.1 | - | 2 | 856 | |||
AK160653.1 | - | 2 | 855 | |||
AK163283.1 | - | 2 | 847 | |||
AK167176.1 | - | 2 | 853 | |||
BB615769.1 | - | 2 | 607 | |||
BC018556.1 | - | 2 | 545 | 3987825 | ||
BC090963.1 | - | 1 | 448 | 30632168 | ||
D17411.1 | - | 2 | 479 | |||
D17412.1 | - | 2 | 479 | |||
D45860.1 | - | 2 | 479 | |||
U09218.1 | - | 2 | 538 |