Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013636.3 | NP_038664.2 | 5 | 1004 | ||
GenBank mRNA | AK076009.1 | - | 5 | 941 | ||
AK088229.1 | - | 5 | 948 | |||
AK147131.1 | - | 5 | 1006 | |||
AK152422.1 | - | 5 | 967 | |||
AK152625.1 | - | 5 | 943 | |||
AK153363.1 | - | 5 | 817 | |||
AK169336.1 | - | 5 | 964 | |||
BC008157.1 | - | 5 | 922 | |||
BC010613.1 | - | 5 | 777 | 3589168 | ||
BC021646.1 | - | 5 | 1005 | 3487479 | ||
BC039972.1 | - | 5 | 937 | |||
BC085496.1 | - | 5 | 943 | 6829867 | ||
D85137.1 | - | 5 | 829 | |||
M27071.1 | - | 5 | 925 | |||
U53456.1 | - | 5 | 921 |