Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163415.1 | NP_001156887.1 | 1413 | |||
NM_011154.2 | NP_035284.2 | 1179 | ||||
GenBank mRNA | AF050165.1 | - | 1176 | |||
AK054122.1 | - | 1552 | ||||
AK143212.1 | - | 1456 | ||||
AK190355.1 | - | 207 | ||||
BC027415.1 | - | 1156 | 4235453 | |||
BC059852.1 | - | 1444 | 6402777 | |||
BC095938.1 | - | 334 | ||||
BC096544.1 | - | 859 | 6834351 |