Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293622.1 | NP_001280551.1 | 6 | 1549 | ||
NM_008913.5 | NP_032939.1 | 6 | 1549 | |||
GenBank mRNA | AK146387.1 | - | 6 | 1493 | ||
AK150393.1 | - | 6 | 908 | |||
AK177425.1 | - | 2 | 291 | |||
AK189167.1 | - | 2 | 291 | |||
AK191293.1 | - | 2 | 291 | |||
AK191345.1 | - | 2 | 291 | |||
AK197466.1 | - | 2 | 290 | |||
AK211949.1 | - | 2 | 290 | |||
AK211995.1 | - | 2 | 291 | |||
AK219430.1 | - | 2 | 290 | |||
BC138612.1 | - | 6 | 822 | 8861632 | ||
J04134.1 | - | 1 | 129 | |||
J05479.1 | - | 6 | 846 |