Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015002.1 | - | 8 | 1152 | ||
AK133393.1 | - | 8 | 1173 | |||
BC046617.1 | - | 8 | 978 | |||
BC141079.1 | - | 8 | 993 | 9055691 | ||
BC145347.1 | - | 7 | 868 | |||
M81475.1 | - | 8 | 1146 |