Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011158.3 | NP_035288.2 | 9, 10 | 1183 | ||
GenBank mRNA | AK041013.1 | - | 9, 10 | 1157 | ||
AK081805.1 | - | 9, 10 | 960 | |||
AK135621.1 | - | 9, 10 | 1183 | |||
AK138963.1 | - | 9, 10 | 1149 | |||
BC033267.1 | - | 5, 6 | 471 |