Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016979.1 | NP_058675.1 | 1 | 359 | ||
GenBank mRNA | AJ238004.1 | - | 1 | 242 | ||
AK034681.1 | - | 1 | 678 | |||
AK036432.1 | - | 1 | 324 | |||
AK037141.1 | - | 1 | 353 | |||
AK039088.1 | - | 1 | 374 | |||
AK081548.1 | - | 1 | 339 | |||
AK139510.1 | - | 1 | 367 | |||
AK149937.1 | - | 1 | 337 | |||
AK150588.1 | - | 1 | 321 | |||
AK152166.1 | - | 1 | 156 | |||
AK154447.1 | - | 1 | 339 | |||
AK169322.1 | - | 1 | 310 | |||
BC006875.1 | - | 1 | 359 | 3596013 |