Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163577.1 | NP_001157049.1 | 19, 20 | 2325 | ||
NM_001163578.1 | NP_001157050.1 | 17, 18 | 2209 | |||
NM_001163581.1 | NP_001157053.1 | 16, 17 | 2119 | |||
NM_001163582.1 | NP_001157054.1 | 16, 17 | 2134 | |||
NM_001163583.1 | NP_001157055.1 | 17, 18 | 2209 | |||
NM_001163584.1 | NP_001157056.1 | 17, 18 | 2209 | |||
NM_008935.2 | NP_032961.2 | 18, 19 | 2298 | |||
GenBank mRNA | AF026269.1 | - | 18, 19 | 2243 | ||
AF039663.1 | - | 19, 20 | 2317 | |||
AF305215.1 | - | 17, 18 | 2154 | |||
AK029921.1 | - | 18, 19 | 2761 | |||
AK030027.1 | - | 18, 19 | 2728 | |||
AY099088.1 | - | 17, 18 | 2154 | |||
AY223521.1 | - | 16, 17 | 2064 | |||
AY223522.1 | - | 16, 17 | 2079 |