Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163577.1 | NP_001157049.1 | 17 | 2131 | ||
NM_001163578.1 | NP_001157050.1 | 15 | 2015 | |||
NM_001163581.1 | NP_001157053.1 | 14 | 1925 | |||
NM_001163582.1 | NP_001157054.1 | 14 | 1940 | |||
NM_001163583.1 | NP_001157055.1 | 15 | 2015 | |||
NM_001163584.1 | NP_001157056.1 | 15 | 2015 | |||
NM_001163585.1 | NP_001157057.1 | 16 | 2042 | |||
NM_008935.2 | NP_032961.2 | 16 | 2104 | |||
GenBank mRNA | AF026269.1 | - | 16 | 2049 | ||
AF039663.1 | - | 17 | 2123 | |||
AF305215.1 | - | 15 | 1960 | |||
AK029921.1 | - | 16 | 2567 | |||
AK030027.1 | - | 16 | 2534 | |||
AY099088.1 | - | 15 | 1960 | |||
AY223521.1 | - | 14 | 1870 | |||
AY223522.1 | - | 14 | 1885 | |||
BC028286.1 | - | 17 | 2141 | 4502359 |