Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PSAP SIRNA Assays ›
Gene Symbol:
Psap
Gene Name:
prosaposin
Gene Aliases:
AI037048, SGP-1
Chromosome Location:
Chr.10: 60277628 - 60302600 on Build GRCm38
UniGene ID:
Mm.277498
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s72250
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Psap
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146120.1
NP_001139592.1
7
846
NM_001146121.1
NP_001139593.1
7
846
NM_001146122.1
NP_001139594.1
7
846
NM_001146123.1
NP_001139595.1
7
846
NM_001146124.1
NP_001139596.1
7
762
NM_011179.3
NP_035309.3
7
846
GenBank mRNA
AK088369.1
-
7
760
AK089998.1
-
7
764
AK093881.1
-
7
760
AK135637.1
-
7
760
AK145607.1
-
7
767
AK145802.1
-
7
761
AK148150.1
-
7
768
AK148152.1
-
7
787
AK148545.1
-
7
762
AK149785.1
-
7
763
AK150331.1
-
7
762
AK150645.1
-
7
760
AK151034.1
-
7
760
AK151173.1
-
7
761
AK151230.1
-
7
760
AK151234.1
-
7
762
AK151251.1
-
7
760
AK151525.1
-
7
762
AK151529.1
-
7
762
AK151686.1
-
7
796
AK151852.1
-
7
762
AK151875.1
-
7
759
AK151891.1
-
7
760
AK151907.1
-
7
762
AK151943.1
-
7
762
AK152117.1
-
7
762
AK152126.1
-
7
760
AK152134.1
-
7
762
AK152203.1
-
7
794
AK152280.1
-
7
760
AK152319.1
-
7
764
AK152355.1
-
7
762
AK152408.1
-
7
765
AK152505.1
-
7
762
AK152557.1
-
7
762
AK152605.1
-
7
762
AK152819.1
-
7
774
AK153404.1
-
7
760
AK159108.1
-
7
790
AK159164.1
-
7
760
AK159167.1
-
7
761
AK159198.1
-
7
760
AK159221.1
-
7
764
AK159244.1
-
7
760
AK159266.1
-
7
760
AK159333.1
-
7
760
AK159341.1
-
7
760
AK159384.1
-
7
760
AK159414.1
-
7
763
AK159427.1
-
7
762
AK159451.1
-
7
760
AK159457.1
-
7
774
AK159463.1
-
7
762
AK159487.1
-
7
760
AK159492.1
-
7
760
AK159510.1
-
7
760
AK159528.1
-
7
760
AK159530.1
-
7
760
AK159589.1
-
7
762
AK159604.1
-
7
760
AK159622.1
-
6
652
AK159634.1
-
7
760
AK159638.1
-
7
761
AK159657.1
-
7
760
AK159664.1
-
7
760
AK159685.1
-
7
760
AK159692.1
-
7
760
AK159709.1
-
7
760
AK159720.1
-
7
760
AK159728.1
-
7
762
AK159840.1
-
7
760
AK159861.1
-
7
760
AK159862.1
-
7
762
AK159877.1
-
7
760
AK159910.1
-
7
760
AK159915.1
-
7
760
AK159936.1
-
7
762
AK159982.1
-
7
760
AK159994.1
-
7
760
AK160015.1
-
7
760
AK160019.1
-
7
762
AK160021.1
-
7
760
AK160024.1
-
7
760
AK160046.1
-
7
760
AK160049.1
-
7
762
AK160073.1
-
7
760
AK160108.1
-
7
760
AK160877.1
-
7
763
AK161313.1
-
7
777
AK166700.1
-
7
760
AK166707.1
-
7
759
AK166744.1
-
7
758
AK166811.1
-
7
760
AK166849.1
-
7
760
AK166879.1
-
7
762
AK166909.1
-
7
760
AK166920.1
-
7
760
AK166946.1
-
7
765
AK166959.1
-
7
760
AK166984.1
-
7
760
AK167014.1
-
7
760
AK167025.1
-
7
760
AK167073.1
-
7
760
AK167099.1
-
7
763
AK167108.1
-
7
760
AK167111.1
-
7
760
AK167147.1
-
7
760
AK167198.1
-
7
767
AK167213.1
-
7
760
AK167237.1
-
7
760
AK167243.1
-
7
761
AK167252.1
-
7
762
AK167418.1
-
7
762
AK167719.1
-
7
762
AK167902.1
-
7
830
AK170416.1
-
7
759
BC036266.1
-
6
626
BC141089.1
-
7
759
9055701
BC145359.1
-
7
759
9053917
S36200.1
-
7
736
U27340.1
-
7
760
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
sphingolipid metabolic process
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
regulation of lipid metabolic process
regulation of MAPK cascade
positive regulation of MAPK cascade
developmental growth
prostate gland growth
epithelial cell differentiation involved in prostate gland development
cellular response to organic substance
negative regulation of hydrogen peroxide-induced cell death
G-protein coupled receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline