Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000597.2 | NP_000588.2 | 3, 4 | 949 | ||
NM_001313990.1 | NP_001300919.1 | 3, 4 | 507 | |||
NM_001313992.1 | NP_001300921.1 | 3, 4 | 826 | |||
NM_001313993.1 | NP_001300922.1 | 3, 4 | 565 | |||
GenBank mRNA | A09809.1 | - | 3, 4 | 948 | ||
BC004312.1 | - | 3, 4 | 923 | 3627826 | ||
BC009902.1 | - | 3, 4 | 925 | 3509749 | ||
BC012769.1 | - | 3, 4 | 923 | 3689990 | ||
BC071967.1 | - | 3, 4 | 936 | 6650239 | ||
M35410.1 | - | 3, 4 | 892 | |||
X16302.1 | - | 3, 4 | 946 |