Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011185.3 | NP_035315.1 | 1 | 112 | ||
GenBank mRNA | AK028265.1 | - | 1 | 134 | ||
AK077520.1 | - | 1 | 113 | |||
AK132161.1 | - | 1 | 113 | |||
AK167647.1 | - | 1 | 134 | |||
BC018351.1 | - | 1 | 113 | 3584538 | ||
U60824.1 | - | 1 | 127 | |||
X80686.1 | - | 1 | 116 |