Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008945.3 | NP_032971.2 | 3 | 519 | ||
GenBank mRNA | AK077448.1 | - | 3 | 496 | ||
AK137044.1 | - | 3 | 495 | |||
AK156624.1 | - | 3 | 495 | |||
AK161348.1 | - | 3 | 495 | |||
AK168265.1 | - | 3 | 495 | |||
AK168340.1 | - | 3 | 495 | |||
AK194462.1 | - | 2 | 224 | |||
AK204169.1 | - | 1 | 37 | |||
BC008241.1 | - | 3 | 474 | 2654421 | ||
CB058553.1 | - | 3 | 519 | |||
U65636.1 | - | 3 | 474 |