Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013831.4 | NP_038859.3 | 1123 | |||
GenBank mRNA | AK132418.1 | - | 1105 | |||
AK154284.1 | - | 1090 | ||||
AK154724.1 | - | 13, 14 | 1129 | |||
AK157994.1 | - | 13, 14 | 1107 | |||
BC057654.1 | - | 1065 | 4460817 | |||
Y18101.1 | - | 13, 14 | 955 |