Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptger2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008964.4 | NP_032990.1 | 2 | 1763 | ||
GenBank mRNA | AK036531.1 | - | 2 | 1681 | ||
AK054132.1 | - | 2 | 1284 | |||
AK086904.1 | - | 2 | 1678 | |||
AK136577.1 | - | 2 | 776 | |||
AK137515.1 | - | 2 | 973 | |||
BC005440.1 | - | 2 | 1271 | 3487087 | ||
D50589.1 | - | 2 | 1054 |
Set Membership: |
G-protein coupled receptor |