Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011200.2 | NP_035330.1 | 5, 6 | 1100 | ||
GenBank mRNA | AK081491.1 | - | 4, 5 | 566 | ||
AK148288.1 | - | 5, 6 | 1096 | |||
AK150506.1 | - | 4, 5 | 730 | |||
AK150556.1 | - | 3, 4 | 1683 | |||
AK150762.1 | - | 3, 4 | 1684 | |||
AK151003.1 | - | 3, 4 | 1684 | |||
AK151533.1 | - | 4, 5 | 730 | |||
BC031734.1 | - | 5, 6 | 1022 | |||
BC055039.1 | - | 5, 6 | 1056 | 6396041 | ||
BC086787.1 | - | 4, 5 | 406 | 30538771 | ||
BC150972.1 | - | 4, 5 | 772 | 9087885 | ||
BC150981.1 | - | 4, 5 | 775 | 9087894 | ||
U84411.1 | - | 5, 6 | 1109 |