Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166388.1 | NP_001159860.1 | 6 | 1895 | ||
NM_001166389.1 | NP_001159861.1 | 6 | 1626 | |||
NM_001166390.1 | NP_001159862.1 | 5 | 727 | |||
NM_008975.3 | NP_033001.2 | 6 | 1695 | |||
GenBank mRNA | AF035645.1 | - | 5 | 676 | ||
AK003954.1 | - | 5 | 433 | |||
AK004562.1 | - | 6 | 1304 | |||
AK014601.1 | - | 6 | 1309 | |||
AK143702.1 | - | 6 | 1347 | |||
AK147489.1 | - | 4 | 6088 | |||
AK172192.1 | - | 6 | 1349 | |||
BC027445.1 | - | 5 | 506 | 4482106 | ||
BC066043.1 | - | 6 | 1895 | 6415021 | ||
CT010215.1 | - | 5 | 461 |