Search Thermo Fisher Scientific
Gene Symbol: Dusp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013642.3 | NP_038670.1 | 3 | 864 | ||
GenBank mRNA | AK149915.1 | - | 3 | 742 | ||
AK150433.1 | - | 3 | 864 | |||
AK152183.1 | - | 3 | 864 | |||
AK152324.1 | - | 3 | 867 | |||
AK152905.1 | - | 2 | 1234 | |||
AK159251.1 | - | 3 | 865 | |||
AK159363.1 | - | 3 | 865 | |||
AK159567.1 | - | 3 | 865 | |||
AK159658.1 | - | 3 | 862 | |||
AK159726.1 | - | 3 | 798 | |||
AK167602.1 | - | 3 | 866 | |||
AK168627.1 | - | 3 | 865 | |||
BC006967.1 | - | 3 | 851 | 3589849 | ||
X61940.1 | - | 3 | 854 |