Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011211.3 | NP_035341.2 | 20 | 1973 | ||
GenBank mRNA | AF326559.1 | - | 2 | 526 | ||
AF326560.1 | - | 2 | 526 | |||
BC086654.1 | - | 11 | 1208 | |||
BC141409.1 | - | 10 | 1152 | 9056022 | ||
D13903.1 | - | 2 | 926 |