Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159525.1 | NP_001152997.1 | 2 | 126 | ||
NM_023041.3 | NP_075528.3 | 4 | 402 | |||
GenBank mRNA | AK012785.1 | - | 4 | 385 | ||
AK028449.1 | - | 2 | 120 | |||
AK029368.1 | - | 4 | 403 | |||
AK075576.1 | - | 4 | 389 | |||
AK170175.1 | - | 4 | 391 | |||
BC012517.1 | - | 4 | 337 | |||
BC019767.1 | - | 4 | 385 | 4502819 | ||
CT010305.1 | - | 4 | 378 | |||
Y09046.1 | - | 4 | 383 |