Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016676.5 | NP_057885.1 | 5 | 985 | ||
GenBank mRNA | AB232604.1 | - | 5 | 434 | ||
AF035646.1 | - | 5 | 675 | |||
AK008725.1 | - | 5 | 975 | |||
AK028320.1 | - | 5 | 956 | |||
AK087964.1 | - | 5 | 947 | |||
AK134988.1 | - | 5 | 905 | |||
AK146357.1 | - | 5 | 986 | |||
AK150319.1 | - | 5 | 945 | |||
AK150384.1 | - | 5 | 639 | |||
AK150791.1 | - | 5 | 935 | |||
AK151473.1 | - | 5 | 639 | |||
AK153315.1 | - | 5 | 945 | |||
AK153325.1 | - | 5 | 935 | |||
AK159383.1 | - | 5 | 950 | |||
AK159845.1 | - | 5 | 950 | |||
AK159988.1 | - | 5 | 944 | |||
AK160960.1 | - | 5 | 940 | |||
AK166010.1 | - | 5 | 959 | |||
AK168320.1 | - | 5 | 945 | |||
AK190935.1 | - | 4 | 287 | |||
AK193085.1 | - | 4 | 268 | |||
AK202554.1 | - | 3 | 278 | |||
AK212462.1 | - | 3 | 219 | |||
BC052735.1 | - | 935 | ||||
BC056374.1 | - | 5 | 936 | 6848840 | ||
CT010394.1 | - | 5 | 434 |