Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008997.3 | NP_033023.1 | 2 | 387 | ||
GenBank mRNA | AB232606.1 | - | 2 | 163 | ||
AK134125.1 | - | 2 | 132 | |||
AK146452.1 | - | 2 | 248 | |||
AK146460.1 | - | 2 | 199 | |||
AK146623.1 | - | 2 | 199 | |||
AK146978.1 | - | 2 | 179 | |||
AK147077.1 | - | 2 | 218 | |||
AK148022.1 | - | 2 | 260 | |||
AK148106.1 | - | 2 | 186 | |||
AK177220.1 | - | 2 | 104 | |||
AK186595.1 | - | 2 | 154 | |||
AK188436.1 | - | 2 | 154 | |||
AK189626.1 | - | 2 | 154 | |||
AK195913.1 | - | 1 | 43 | |||
BC054753.1 | - | 2 | 203 | 6411651 | ||
BC085270.1 | - | 2 | 196 | 6478784 | ||
BC157916.1 | - | 2 | 216 | 9007344 | ||
BU705914.1 | - | 2 | 387 | |||
BY275360.1 | - | 2 | 382 | |||
KJ650201.1 | - | 2 | 163 | |||
L26528.1 | - | 2 | 198 |