Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008997.3 | NP_033023.1 | 5 | 769 | ||
GenBank mRNA | AB232606.1 | - | 5 | 545 | ||
AK134125.1 | - | 5 | 514 | |||
AK146452.1 | - | 5 | 630 | |||
AK146460.1 | - | 5 | 581 | |||
AK146623.1 | - | 5 | 581 | |||
AK146978.1 | - | 5 | 561 | |||
AK147077.1 | - | 5 | 600 | |||
AK148022.1 | - | 5 | 642 | |||
AK148106.1 | - | 5 | 568 | |||
AK188721.1 | - | 3 | 218 | |||
AK203890.1 | - | 3 | 243 | |||
AK205000.1 | - | 3 | 243 | |||
BC054753.1 | - | 5 | 585 | 6411651 | ||
BC085270.1 | - | 5 | 578 | 6478784 | ||
BC157916.1 | - | 5 | 598 | 9007344 | ||
KJ650201.1 | - | 5 | 545 | |||
L26528.1 | - | 5 | 580 |