Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177411.4 | NP_803130.1 | 4 | 560 | ||
GenBank mRNA | AB232594.1 | - | 3 | 421 | ||
AK081251.1 | - | 4 | 556 | |||
AK083034.1 | - | 4 | 549 | |||
AK144551.1 | - | 4 | 682 | |||
AK218600.1 | - | 1 | 22 | |||
BC137574.1 | - | 3 | 472 | 8860593 | ||
BC137575.1 | - | 3 | 472 | 8860594 | ||
BY727964.1 | - | 4 | 561 | |||
X84239.1 | - | 3 | 429 |