Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252562.1 | NP_001239491.1 | 2 | 108 | ||
NM_009014.3 | NP_033040.2 | 2 | 126 | |||
GenBank mRNA | AK015771.1 | - | 2 | 108 | ||
AK032913.1 | - | 2 | 127 | |||
U92068.1 | - | 1 | 68 |