Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009022.4 | NP_033048.2 | 13 | 1567 | ||
GenBank mRNA | AK078553.1 | - | 13 | 1508 | ||
BC075704.1 | - | 13 | 1534 | 30471325 | ||
X99273.1 | - | 13 | 1512 |