Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009029.2 | NP_033055.2 | 17 | 1687 | ||
GenBank mRNA | AK132875.1 | - | 6 | 433 | ||
AK141028.1 | - | 17 | 1719 | |||
AK146467.1 | - | 12 | 1018 | |||
BC096525.1 | - | 17 | 1687 | 6832808 | ||
DQ400415.1 | - | 17 | 1537 | |||
M26391.1 | - | 17 | 1635 |