Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009029.2 | NP_033055.2 | 20 | 2109 | ||
GenBank mRNA | AK132875.1 | - | 9 | 855 | ||
AK141028.1 | - | 20 | 2141 | |||
AK146467.1 | - | 15 | 1440 | |||
BC096525.1 | - | 20 | 2109 | 6832808 | ||
DQ400415.1 | - | 20 | 1959 | |||
M26391.1 | - | 20 | 2057 |