Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009029.2 | NP_033055.2 | 11, 12 | 1267 | ||
GenBank mRNA | AK011246.1 | - | 11, 12 | 1267 | ||
AK132875.1 | - | 13 | ||||
AK141028.1 | - | 11, 12 | 1299 | |||
AK146467.1 | - | 6, 7 | 598 | |||
AK204056.1 | - | 1, 2 | 35 | |||
BC096525.1 | - | 11, 12 | 1267 | 6832808 | ||
DQ400415.1 | - | 11, 12 | 1117 | |||
M26391.1 | - | 11, 12 | 1215 |