Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009053.2 | NP_033079.1 | 5 | 636 | ||
GenBank mRNA | AF015770.1 | - | 5 | 594 | ||
AK004573.1 | - | 4 | 908 | |||
AK183109.1 | - | 2 | 99 | |||
AK207192.1 | - | 4 | 241 | |||
AK212815.1 | - | 2 | 142 | |||
AK217572.1 | - | 1 | 28 | |||
BC066023.1 | - | 5 | 636 | 6829176 | ||
U94350.1 | - | 5 | 629 |