Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165934.1 | NP_001159406.1 | 13 | 1083 | ||
NM_011268.2 | NP_035398.2 | 13 | 1083 | |||
GenBank mRNA | AF011358.1 | - | 13 | 968 | ||
AF125046.1 | - | 13 | 1039 | |||
AK138159.1 | - | 13 | 993 | |||
BC129898.1 | - | 5 | 361 | 40111530 | ||
BC129899.1 | - | 13 | 949 | 40111531 |