Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rpgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177950.1 | NP_001171421.1 | 18 | 3068 | ||
NM_001177952.1 | NP_001171423.1 | 17 | 2183 | |||
NM_011285.2 | NP_035415.1 | 17 | 2189 | |||
GenBank mRNA | AF044677.1 | - | 17 | 2107 | ||
AJ238396.1 | - | 18 | 2980 | |||
AK136298.1 | - | 17 | 2089 | |||
AK138977.1 | - | 17 | 2184 |