Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011294.3 | NP_035424.1 | 4 | 332 | ||
GenBank mRNA | AK030954.1 | - | 1 | 1283 | ||
AK048371.1 | - | 4 | 332 | |||
AK049260.1 | - | 4 | 332 | |||
AK132021.1 | - | 4 | 331 | |||
AK146335.1 | - | 4 | 331 | |||
AK151734.1 | - | 4 | 332 | |||
AK154114.1 | - | 4 | 331 | |||
BC010967.1 | - | 4 | 330 | 4018261 | ||
J03750.1 | - | 4 | 316 |