Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009099.2 | NP_033125.2 | 3, 4 | 755 | ||
GenBank mRNA | AF220014.1 | - | 3, 4 | 672 | ||
AK134144.1 | - | 3, 4 | 724 | |||
AK137506.1 | - | 3, 4 | 756 | |||
AK138249.1 | - | 4, 5 | 758 | |||
BC005447.1 | - | 3, 4 | 689 | 3493256 | ||
J03776.1 | - | 3, 4 | 672 |