Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000880.3 | NP_000871.1 | 2 | 711 | ||
NM_001199886.1 | NP_001186815.1 | 2 | 711 | |||
NM_001199887.1 | NP_001186816.1 | 2 | 711 | |||
NM_001199888.1 | NP_001186817.1 | 2 | 711 | |||
XM_011517522.2 | XP_011515824.1 | 2 | 651 | |||
XM_011517523.2 | XP_011515825.1 | 2 | 651 | |||
GenBank mRNA | AB102879.1 | - | 1 | 110 | ||
AB102885.1 | - | 1 | 110 | |||
AB102893.1 | - | 1 | 110 | |||
AK226000.1 | - | 2 | 652 | |||
BC047698.1 | - | 2 | 699 | 5748841 | ||
CD013923.1 | - | 2 | 116 | |||
GQ919059.1 | - | 1 | 35 | |||
J04156.1 | - | 2 | 494 |