Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011304.3 | NP_035434.1 | 11 | 1121 | ||
GenBank mRNA | AB013912.1 | - | 11 | 952 | ||
AK135404.1 | - | 11 | 935 | |||
AK144156.1 | - | 11 | 943 | |||
AK159112.1 | - | 11 | 1123 | |||
AK189162.1 | - | 3 | 177 | |||
AK191656.1 | - | 3 | 177 | |||
AK210146.1 | - | 3 | 177 | |||
AK212334.1 | - | 3 | 177 | |||
AK212947.1 | - | 3 | 177 | |||
BC072585.1 | - | 1620 | ||||
BC115809.1 | - | 11 | 939 | 40105495 | ||
BC116693.1 | - | 11 | 938 | 40105493 |