Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159731.1 | NP_001153203.1 | 2 | 301 | ||
NM_009107.3 | NP_033133.1 | 3 | 757 | |||
GenBank mRNA | BC058401.1 | - | 3 | 739 | 5707723 | |
BM947610.1 | - | 3 | 483 | |||
BY769777.1 | - | 2 | 302 | |||
G49237.1 | - | 3 | 489 | |||
M84819.1 | - | 3 | 402 | |||
S62948.1 | - | 3 | 601 | |||
S62952.1 | - | 2 | 257 | |||
X66225.1 | - | 3 | 489 |