Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163504.1 | NP_001156976.1 | 5 | 897 | ||
NM_001163700.1 | NP_001157172.1 | 4 | 693 | |||
NM_009108.2 | NP_033134.2 | 4 | 681 | |||
GenBank mRNA | AK002513.1 | - | 4 | 679 | ||
AK132255.1 | - | 4 | 673 | |||
BC015261.1 | - | 4 | 652 | 4241705 | ||
U09416.1 | - | 4 | 650 | |||
U09417.1 | - | 5 | 861 | |||
U09418.1 | - | 5 | 858 |