Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199304.1 | NP_001186233.1 | 6 | 2123 | ||
NM_001199305.1 | NP_001186234.1 | 7 | 2248 | |||
NM_009124.6 | NP_033150.2 | 7 | 2248 | |||
GenBank mRNA | AK048268.1 | - | 7 | 2235 | ||
AK169931.1 | - | 7 | 2244 | |||
BC058178.1 | - | 6 | 2089 | 5326559 | ||
X83542.1 | - | 6 | 1989 |