Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011330.3 | NP_035460.1 | 3 | 815 | ||
GenBank mRNA | AK009307.1 | - | 3 | 799 | ||
AK010146.1 | - | 3 | 726 | |||
BC027521.1 | - | 3 | 715 | 1527856 | ||
U26426.1 | - | 3 | 714 | |||
U40672.1 | - | 3 | 728 |