Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011330.3 | NP_035460.1 | 2 | 286 | ||
GenBank mRNA | AF128205.1 | - | 2 | 172 | ||
AF128206.1 | - | 2 | 172 | |||
AF128207.1 | - | 2 | 172 | |||
AF128208.1 | - | 2 | 172 | |||
AF128209.1 | - | 2 | 172 | |||
AK009307.1 | - | 2 | 269 | |||
AK010146.1 | - | 2 | 197 | |||
AW986913.1 | - | 2 | 286 | |||
BC027521.1 | - | 2 | 186 | 1527856 | ||
U26426.1 | - | 2 | 185 | |||
U40672.1 | - | 2 | 198 |