Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009138.3 | NP_033164.1 | 4 | 352 | ||
GenBank mRNA | AJ249480.1 | - | 4 | 325 | ||
AK131875.1 | - | 4 | 353 | |||
AK132700.1 | - | 4 | 377 | |||
AK134138.1 | - | 4 | 337 | |||
BC117033.1 | - | 3 | 285 | 40126352 | ||
BC117037.1 | - | 3 | 285 | 40126356 | ||
DQ158256.1 | - | 4 | 338 | |||
U86357.1 | - | 3 | 234 |