Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sord | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146126.4 | NP_666238.1 | 7 | 790 | ||
GenBank mRNA | AK004692.1 | - | 7 | 814 | ||
AK015059.1 | - | 7 | 791 | |||
AK152169.1 | - | 7 | 839 | |||
AK166988.1 | - | 7 | 790 | |||
AK166996.1 | - | 7 | 819 | |||
AK184602.1 | - | 1 | 120 | |||
BC024124.1 | - | 7 | 805 | 5099395 | ||
BC030875.1 | - | 7 | 779 | 4237164 | ||
BC039957.1 | - | 7 | 790 | |||
BC092291.1 | - | 7 | 793 | 5097505 | ||
U27014.1 | - | 790 |