Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039089.1 | NP_001034178.1 | 12 | 1313 | ||
NM_011344.2 | NP_035474.1 | 12 | 1163 | |||
GenBank mRNA | AF063095.1 | - | 12 | 1112 | ||
AK005023.2 | - | 12 | 1286 | |||
AK146022.1 | - | 12 | 1287 | |||
AK160936.1 | - | 12 | 1121 | |||
AK161546.1 | - | 12 | 1287 | |||
AK220502.1 | - | 12 | 1313 | |||
BC026816.1 | - | 12 | 1135 | 4457637 | ||
BC049959.1 | - | 7 | 525 |