Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243072.1 | NP_001230001.1 | 2 | 448 | ||
NM_001243073.1 | NP_001230002.1 | 2 | 288 | |||
NM_009152.4 | NP_033178.2 | 1 | 716 | |||
GenBank mRNA | AK053115.1 | - | 1 | 449 | ||
AK140817.1 | - | 2 | 289 | |||
AK164242.1 | - | 4 | 491 | |||
BC057588.1 | - | 1 | 363 | 6416990 | ||
BC090844.1 | - | 2 | 448 | 30532393 | ||
CN460380.1 | - | 1 | 645 | |||
D85028.2 | - | 1 | 718 | |||
L41541.1 | - | 1 | 67 | |||
X85993.1 | - | 1 | 177 |